missing translation for 'onlineSavingsMsg'
Learn More

Zebrafish Gut Absorptive Cell Epitopes Antibody (FIS 4E8/1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£150.00 - £302.00

Specifications

Antigen Zebrafish Gut Absorptive Cell Epitopes
Clone FIS 4E8/1
Concentration 1 mg/mL
Dilution Western Blot 1:200, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18652829
View Documents
Novus Biologicals
NBP2-50374-0.025ml
0.025 mL
£150.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18638228
View Documents
Novus Biologicals
NBP2-50374
0.1 mL
£302.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zebrafish Gut Absorptive Cell Epitopes Monoclonal antibody specifically detects Zebrafish Gut Absorptive Cell Epitopes in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications

Specifications

Zebrafish Gut Absorptive Cell Epitopes
1 mg/mL
Western Blot, Immunohistochemistry, Immunoprecipitation
Unconjugated
Mouse
Cell Cycle and Replication, Cellular Signaling, Developmental Biology, Epigenetics, Epitope Tags
PBS
IgG1
Protein A purified
FIS 4E8/1
Western Blot 1:200, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Monoclonal
Purified
RUO
Zebrafish
Lysate of zebrafish intestines
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.