missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Gut Absorptive Cell Epitopes Antibody (FIS 4E8/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50374
This item is not returnable.
View return policy
Description
Zebrafish Gut Absorptive Cell Epitopes Monoclonal antibody specifically detects Zebrafish Gut Absorptive Cell Epitopes in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| Zebrafish Gut Absorptive Cell Epitopes | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:200, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| FIS 4E8/1 | |
| Unconjugated | |
| PBS | |
| Lysate of zebrafish intestines | |
| 0.1 mL | |
| Cell Cycle and Replication, Cellular Signaling, Developmental Biology, Epigenetics, Epitope Tags | |
| Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction