missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Ephrin-B2 Antibody, R&D Systems™

Goat Polyclonal Antibody
£170.00 - £451.00
Specifications
| Antigen | Ephrin-B2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18726417
![]() |
R&D Systems
AF1088-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18031952
![]() |
R&D Systems
AF1088 |
100 μg |
£451.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ephrin-B2 Polyclonal specifically detects Ephrin-B2 in Zebrafish samples. It is validated for Western Blot.Specifications
| Ephrin-B2 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Zebrafish | |
| EFNB2, ELF-2, ephrin B2, EphrinB2, EPLG5, Htk-L, HTKL, LERK5, LERK-5, NLERK-1 | |
| efnb2a | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| O73874 | |
| 1948 | |
| Mouse myeloma cell line NS0-derived recombinant zebrafish Ephrin-B2 Leu25-Ala222 Accession # O73874 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects zebrafish Ephrin-B2 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) Ephrin-B2 is observed and less than 1% cross-reactivity with recombinant human (rh) Ephrin-A4, rmEphrin-A2, rmEphrin-A1, rhEphrin-B3, rmEphrin-B1, rhEphrin-A3, rhEphrin-A5, and rmEphrin-A4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title