missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Ephrin-B2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1088-SP
This item is not returnable.
View return policy
Description
Ephrin-B2 Polyclonal specifically detects Ephrin-B2 in Zebrafish samples. It is validated for Western Blot.
Specifications
| Ephrin-B2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EFNB2, ELF-2, ephrin B2, EphrinB2, EPLG5, Htk-L, HTKL, LERK5, LERK-5, NLERK-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1948 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| O73874 | |
| efnb2a | |
| Mouse myeloma cell line NS0-derived recombinant zebrafish Ephrin-B2 Leu25-Ala222 Accession # O73874 | |
| 25 μg | |
| Primary | |
| Detects zebrafish Ephrin-B2 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) Ephrin-B2 is observed and less than 1% cross-reactivity with recombinant human (rh) Ephrin-A4, rmEphrin-A2, rmEphrin-A1, rhEphrin-B3, rmEphrin-B1, rhEphrin-A3, rhEphrin-A5, and rmEphrin-A4 is observed. | |
| Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction