missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZEB1 Mouse anti-Human, Clone: 1H1F1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ZEB1 |
|---|---|
| Clone | 1H1F1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16888005
|
Proteintech
66279-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16878005
|
Proteintech
66279-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
THOC3 is part of the TREX (transcription/export) complex, which includes THO2, HPR1, ALY, and UAP56.TEX1 is part of the TREX (transcription/export) complex, which includes THO2 (MIM 300395), HPR1 (MIM 606930), ALY (MIM 604171), and UAP56 (MIM 606390).Specifications
| ZEB1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| AREB6, BZP, DELTA EF1, delta EF1, Negative regulator of IL2, NIL 2 A, NIL 2 A zinc finger protein, NIL 2A, NIL2A, TCF 8, TCF8, Transcription factor 8, ZEB, ZEB1, ZFHEP, ZFHX1A | |
| ZEB1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1H1F1 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| P37275 | |
| 6935 | |
| ZEB1 Fusion Protein Ag17118 | |
| Primary | |
| -20°C | |
| ZEB1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title