missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZEB1 Mouse anti-Human, Clone: 1H1F1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66279-1-IG-20UL
This item is not returnable.
View return policy
Description
THOC3 is part of the TREX (transcription/export) complex, which includes THO2, HPR1, ALY, and UAP56.TEX1 is part of the TREX (transcription/export) complex, which includes THO2 (MIM 300395), HPR1 (MIM 606930), ALY (MIM 604171), and UAP56 (MIM 606390).Specifications
| ZEB1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P37275 | |
| ZEB1 | |
| ZEB1 Fusion Protein Ag17118 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1H1F1 | |
| Unconjugated | |
| ZEB1 | |
| AREB6, BZP, DELTA EF1, delta EF1, Negative regulator of IL2, NIL 2 A, NIL 2 A zinc finger protein, NIL 2A, NIL2A, TCF 8, TCF8, Transcription factor 8, ZEB, ZEB1, ZFHEP, ZFHX1A | |
| Mouse | |
| Protein G | |
| RUO | |
| 6935 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction