missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC15 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ZDHHC15 |
|---|---|
| Concentration | 0.15 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16891225
|
Proteintech
21627-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16881225
|
Proteintech
21627-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene belongs to the DHHC palmitoyltransferase family. Mutations in this gene are associated with mental retardation X-linked type 91 (MRX91). Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| ZDHHC15 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q96MV8 | |
| 158866 | |
| ZDHHC15 Fusion Protein Ag16152 | |
| Primary | |
| -20°C | |
| ZDHHC15 |
| 0.15 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DHHC 15, MRX91, Palmitoyltransferase ZDHHC15, ZDHHC15 | |
| ZDHHC15 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title