Learn More
ZDHHC15 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21627-1-AP-20UL
 View more versions of this product
Product Code. 16891225
  • £135.00 / 20µL
Description
The protein encoded by this gene belongs to the DHHC palmitoyltransferase family. Mutations in this gene are associated with mental retardation X-linked type 91 (MRX91). Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
ZDHHC15
Polyclonal
Unconjugated
ZDHHC15
DHHC 15, MRX91, Palmitoyltransferase ZDHHC15, ZDHHC15
Rabbit
Antigen Affinity Chromatography
RUO
158866
-20°C
Liquid
Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96MV8
ZDHHC15
ZDHHC15 Fusion Protein Ag16152
20 μL