missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£361.00
Specifications
| Antigen | ZDHHC13 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
ZDHHC13 Polyclonal specifically detects ZDHHC13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ZDHHC13 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Q8IUH4-3 | |
| 54503 | |
| Synthetic peptides corresponding to ZDHHC13(zinc finger, DHHC-type containing 13) The peptide sequence was selected from the N terminal of ZDHHC13. Peptide sequence MVILLLQHGADPTLIDGEGFSSIHLAVLFQHMPIIAYLISKGQSVNMTDV. | |
| Primary |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| DHHC-13, EC 2.3.1, EC 2.3.1.-, FLJ10852, FLJ10941, HIP14LHIP14-related protein, HIP3RP, huntingtin interacting protein HIP3RP, Huntingtin-interacting protein 14-related protein, Huntingtin-interacting protein HIP3RP, MGC64994, palmitoyltransferase ZDHHC13, probable palmitoyltransferase ZDHHC13, Putative MAPK-activating protein PM03, Putative NF-kappa-B-activating protein 209, Zinc finger DHHC domain-containing protein 13, zinc finger, DHHC domain containing 13, zinc finger, DHHC-type containing 13 | |
| ZDHHC13 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title