missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59026
This item is not returnable.
View return policy
Description
ZDHHC13 Polyclonal specifically detects ZDHHC13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ZDHHC13 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DHHC-13, EC 2.3.1, EC 2.3.1.-, FLJ10852, FLJ10941, HIP14LHIP14-related protein, HIP3RP, huntingtin interacting protein HIP3RP, Huntingtin-interacting protein 14-related protein, Huntingtin-interacting protein HIP3RP, MGC64994, palmitoyltransferase ZDHHC13, probable palmitoyltransferase ZDHHC13, Putative MAPK-activating protein PM03, Putative NF-kappa-B-activating protein 209, Zinc finger DHHC domain-containing protein 13, zinc finger, DHHC domain containing 13, zinc finger, DHHC-type containing 13 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Mouse: 100%; Rat: 100%; Canine: 92%; Equine: 92%; Rabbit: 92%; Chicken: 91%; Bovine: 85%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q8IUH4-3 | |
| ZDHHC13 | |
| Synthetic peptides corresponding to ZDHHC13(zinc finger, DHHC-type containing 13) The peptide sequence was selected from the N terminal of ZDHHC13. Peptide sequence MVILLLQHGADPTLIDGEGFSSIHLAVLFQHMPIIAYLISKGQSVNMTDV. | |
| 100 μL | |
| Signal Transduction | |
| 54503 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction