missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC11/ZDHHC11B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79327
This item is not returnable.
View return policy
Description
ZDHHC11 Polyclonal specifically detects ZDHHC11 in Rat samples. It is validated for Western Blot.
Specifications
| ZDHHC11 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DHHC-11, EC 2.3.1.-, FLJ13153, Zinc finger DHHC domain-containing protein 11, Zinc finger protein 399, zinc finger, DHHC domain containing 11, zinc finger, DHHC-type containing 11, ZNF399probable palmitoyltransferase ZDHHC11 | |
| Rabbit | |
| 42 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Rat: 92%. | |
| Human, Mouse, Rat, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P0C7U3 | |
| ZDHHC11 | |
| The immunogen for this antibody is Zdhhc11 (NP_001034431). Peptide sequence TIDPADTNVRLKKDYLEPVPTFDRSKHAHVIQNQYCHLCEVTVSKKAKHC. | |
| Affinity purified | |
| RUO | |
| 79844 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction