missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB46 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£156.00 - £301.00
Specifications
| Antigen | ZBTB46 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | ELISA 1:100-1:1000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18120613
|
Novus Biologicals
NBP2-27211SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18114645
|
Novus Biologicals
NBP2-27211 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZBTB46 Polyclonal specifically detects ZBTB46 in Human samples. It is validated for ELISA.Specifications
| ZBTB46 | |
| ELISA 1:100-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 140685 | |
| Amino acids 62-87 of human RINZF protein were used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| BTB (POZ) domain containing 4, BTB/POZ domain-containing protein 4, BTBD4, dJ583P15.7, dJ583P15.8, FLJ13502, RINZF, zinc finger and BTB domain containing 46, zinc finger and BTB domain-containing protein 46, Zinc finger protein 340, ZNF340 | |
| ZBTB46 | |
| IgG | |
| Protein G purified | |
| The NCBI accession number forRINZF is Q96DT7. The amino acid sequence used as immunogen is 100% homologous in human and chimp and 88% homologous in mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title