missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB46 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-27211SS
This item is not returnable.
View return policy
Description
ZBTB46 Polyclonal specifically detects ZBTB46 in Human samples. It is validated for ELISA.
Specifications
| ZBTB46 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| ZBTB46 | |
| Amino acids 62-87 of human RINZF protein were used as the immunogen. | |
| 0.025 mg | |
| Primary | |
| The NCBI accession number forRINZF is Q96DT7. The amino acid sequence used as immunogen is 100% homologous in human and chimp and 88% homologous in mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 0.5 mg/mL | |
| ELISA 1:100-1:1000 | |
| BTB (POZ) domain containing 4, BTB/POZ domain-containing protein 4, BTBD4, dJ583P15.7, dJ583P15.8, FLJ13502, RINZF, zinc finger and BTB domain containing 46, zinc finger and BTB domain-containing protein 46, Zinc finger protein 340, ZNF340 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 140685 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction