missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBED3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£141.00
Specifications
| Antigen | ZBED3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
ZBED3 Polyclonal specifically detects ZBED3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| ZBED3 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_115743 | |
| 84327 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human ZBED3. The immunogen is located within the first 50 amino acids of ZBED3. Amino Acid Squence: GPAPTPTPPGRLGAPYS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 28 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| BED domain containing 3, MGC15435,2610005H11Rik, zinc finger, BED-type containing 3 | |
| ZBED3 | |
| IgG | |
| Affinity Purified | |
| This antibody is predicted to have no cross-reactivity to ZBED1 or ZBED2. |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit