missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBED3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76326-0.025mg
This item is not returnable.
View return policy
Description
ZBED3 Polyclonal specifically detects ZBED3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| ZBED3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| BED domain containing 3, MGC15435,2610005H11Rik, zinc finger, BED-type containing 3 | |
| Rabbit | |
| 28 kDa | |
| 0.025 mg | |
| Primary | |
| This antibody is predicted to have no cross-reactivity to ZBED1 or ZBED2. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 | |
| NP_115743 | |
| ZBED3 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human ZBED3. The immunogen is located within the first 50 amino acids of ZBED3. Amino Acid Squence: GPAPTPTPPGRLGAPYS | |
| Affinity Purified | |
| RUO | |
| 84327 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction