missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Z-Val-Val-Nle Diazomethyl Ketone

Product Code. 15436449
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436449 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15436449 Supplier Thermo Scientific Chemicals Supplier No. J64544.MF
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 155026-49-6 | C25H37N5O5, C25H37N5O5 | 487.60 g/mol

Z-Val-Val-Nle diazomethyl ketone is used as an irreversible cathepsin inhibitor that is 380 times more effective in inactivating cathepsin S than L. It is often abbreviated as DMK. Peptidyl diazomethyl ketones are specific inactivators of thiol proteinases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Description White to Off-white
Content And Storage -30°C to -10°C
For Use With (Application) Inactivates thiol proteinases; an irreversible cathepsin inhibitor
CAS 155026-49-6
Molecular Formula C25H37N5O5
Formula Weight 487.60g/mol
Synonym C-3890; Z-Val-Val-Nle-DMK
Product Type Signal Transduction Reagent
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.