missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) trifluoromethanesulfonate hydrate

Product Code. 11303878
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303878 2 g 2g
11383868 10 g 10g
11393868 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11303878 Supplier Thermo Scientific Alfa Aesar Supplier No. 040315.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34629-25-9 | C3F9O9S3Y | 536.10 g/mol

It is employed in preparation of superconducting thin films. It is used in chemical research & as a intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34629-25-9
Molecular Formula C3F9O9S3Y
Molecular Weight (g/mol) 536.10
MDL Number MFCD00209623
InChI Key JPJIEXKLJOWQQK-UHFFFAOYSA-K
Synonym yttrium iii trifluoromethanesulfonate hydrate, acmc-20aj8x, 3cf3o3s.y.h2o, yttrium 3+ hydrate tritriflate, yttrium trifluoromethanesulfonate-water 1/3/1, methanesulfonic acid,1,1,1-trifluoro-, yttrium 3+ salt, hydrate 3:1:?
PubChem CID 57349102
SMILES [Y+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Physical Form Powder
Quantity 2 g
IUPAC Name yttrium(3+) tritrifluoromethanesulfonate
Formula Weight 536.10 (Anhydrous)
Chemical Name or Material Yttrium(III) trifluoromethanesulfonate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.