accessibility menu, dialog, popup

UserName

FFFSOOOFFFSOOOYFFFSOOO

CAS RN 34629-25-9

IUPAC Name: yttrium(3+) hydrate tritrifluoromethanesulfonate
Synonyms: yttrium(3+) hydrate tritriflate
Molecular Weight (g/mol): 554.11
Molecular Formula: C3H2F9O10S3Y
InChi Key: OYPZNLILUFMJEI-UHFFFAOYSA-K
SMILES: O.[Y+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results