missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YSI™ ProPlus Galvanic DO/pH/ORP/Conductivity Quatro Kit with 4m Cable
Description
Includes
Pro Plus handheld, 4m Quatro cable with galvanic DO, pH/ORP and conductivity sensors, 5913 cap membrane kit (1.25 mil PE), soft-sided carrying case
Pro Plus Meters
- Rubber, over-molded IP67 waterproof case
- Rugged cables, sensors, and military-spec connectors
- Drop-tested 1m at all angles
- Scratch-resistant lens; viewable with polarized glasses
- Instrument floats
- Backlit display and keypad
- 50 data-set memory ensures data integrity
- USB connection powers the instrument
- Auto-buffer recognition; up to 6-point cal
- Detailed GLP data
- Power: 2 alkaline C-cells provide 400 continuous hours at ambient temperature without backlight
- Free, Data Manager desktop software and ProComm II saddle
Certifications
CE, WEEE
Compliance
RoHS
Warranty
3-year instrument warranty; 2-year cable warranty
Specifications
Specifications
| Includes | Pro Plus handheld, 4m Quatro cable with galvanic DO, pH/ORP and conductivity sensors, 5913 cap membrane kit (1.25 mil PE), soft-sided carrying case |
| Calibration | Up to 6-point cal |
| Display Type | Backlit |
| Backlight Option | Yes |
| Memory | 50 data-sets |
| Data Management Details | Free, Data Manager desktop software and ProComm II saddle |
| Certifications/Compliance | RoHS, CE, WEEE |
| IP Rating | IP67 |
| Warranty | 3 Year Instrument, 2 Year Cable |
| Model | ProPlus |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?