missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YSI™ Buffer Solutions for pH Electrodes
Designed and manufactured to the highest quality standards
Brand: YSI™ 003821
Code : 04
Additional Details : Weight : 1.00000kg
Description
- Calibration buffer
- For use with YSI 60, YSI 556, Ecosense™ pH10, Ecosense pH100, 6-Series sonde meters
- Color coded: pH 4 (red), pH 7 (yellow), pH 10 (blue)
Specifications
Red | |
Calibration Buffer | |
4 |