missing translation for 'onlineSavingsMsg'
Learn More

YSI™ Buffer Solutions for pH Electrodes

Product Code. p-4400304
Click to view available options
Color:
Blue
Yellow
pH Range:
7
10
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 12963347

Brand: YSI™ 003823

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed and manufactured to the highest quality standards

  • Calibration buffer
  • For use with YSI 60, YSI 556, Ecosense™ pH10, Ecosense pH100, 6-Series sonde meters
  • Color coded: pH 4 (red), pH 7 (yellow), pH 10 (blue)

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Type Calibration Buffer
pH Range 10
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.