missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YPEL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15403-1-AP-20UL
This item is not returnable.
View return policy
Description
15403-1-AP was raised against full length of YPEL3 protein (1-119 aa); it can recognize YPEL1-4.
YPEL3 (yippee-like 3) belongs to a family of five yippee-like proteins, all of which localize to the centrosome or mitotic spindle and are widely expressed in both adult and fetal tissue. This localization plus the fact that the family of human YPEL proteins share a high degree of sequence homology across species suggests that these proteins may have a conserved function involved in cell division. YPEL3 is a p53-regulated gene whose expression is induced by DNA damage and in turn induces cellular senescence. It appears to function as a tumor suppressor as it is downregulated in colon and breast tumors.Specifications
| YPEL3 | |
| Polyclonal | |
| Unconjugated | |
| YPEL3 | |
| Protein yippee like 3, yippee like 3 (Drosophila), YPEL3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 293491, 66090, 83719 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P61236, P61237 | |
| Ypel3 | |
| YPEL3 Fusion Protein Ag7626 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction