missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Yersinia Selective Agar

Product Code. p-4523654
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12972608

Brand: Thermo Scientific™ PO0287A

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Isolate and enumerate Yersinia enterocolitica from clinical specimens and food samples on this prepared media plate after 18 to 24 hours incubation at 32°C.

Isolate and enumerate Yersinia enterocolitica from clinical specimens and food samples after 18 to 24 hours using Thermo Scientific™ Yersinia Selective Agar (CIN Agar).

Specifically developed for the optimum growth and recovery of Yersinia enterocolitica, the formulation includes cefsulodin, Irgasan™ and novobiocin. The typical colonies of Yersinia enterocolitica will develop as a red bullseye surrounded by a transparent border and will vary considerably among serotypes in colony size, smoothness and the ratio of the border to center diameter. Most other organisms that are capable of growing will produce larger colonies (>2mm in diameter) with diffuse pinkish centers and opaque outer zones. Serratia liquefaciens, Citrobacter freundii and Enterobacter agglomerans may give a colonial morphology resembling Yersinia enterocolitica. These organisms can be differentiated from Yersinia enterocolitica by biochemical tests.

Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery we recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Tested in accordance with CEN ISO/TS 11133-2.The formulation of this medium conforms to ISO 10273.
Color Pink / Red, clear
Content And Storage 2°C to 10°C
Description Yersinia Selective Agar
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 19.5 g ± 1.0 g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.