missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Yellow Universal Pipettor Tips

Product Code. 11817832 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
200 μL
Unit Size:
Pack of 960

Product Code. 11817832

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Make pipetting much simpler and safe for even the most sensitive applications. Yellow Universal Pipettor Tips are perfectly fit for manual or electronic, single or multichannel pipets.

  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types

Specifications

Volume (Metric) 200 μL
Filtered Filtered
Color Yellow
Autoclavable Autoclavable
For Use With (Equipment) Universal Fit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.