missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Plastic Radiation Hazard Trays
Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
46 x 26cm
57 x 54cm
113 x 54cm
Unit Size:
Each
Description
- Available in general purpose, biohazard and radiation safety formats
- Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
- All trays supplied with one free liner when purchased
- Additional environmentally friendly APET liners are provided in packs of 25
Specifications
Specifications
| Dimensions (L x W) | 57 x 54cm |
| Color | Yellow |
| Material | Plastic |
| Type | Radiation Hazard Tray |
| For Use With | Safe containment of spillages, lab bench protection |
| Includes | Liner |
| No. per Pack | 1 Each |
| Shape | Rectangular |
Product Content Correction
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur