missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Yeast Extract Glucose Chloramphenicol Medium (YGC-Medium)

Product Code. 10240973
Change view
Click to view available options
Description:
Yeast Extract Glucose Chloramphenicol Medium
Quantity:
10/Pk.
Fill Volume:
17 mL
Fill Weight:
17g ± 5%
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity Fill Volume Fill Weight Unit Size
10240973 Yeast Extract Glucose Chloramphenicol Medium 10/Pk. 17 mL 17g ± 5% Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10240973 Supplier Thermo Scientific™ Supplier No. PO5032A
  • ONLINE EXCLUSIVE PRICE
    £18.80
    £18.41 / Pack of 10
    Save £0.39
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and enumerate yeasts and molds in food products with this selective medium.

Isolate and enumerate yeasts and molds from food samples using Thermo Scientific™ Yeast Extract Glucose Chloramphenicol Medium. Chloramphenicol in the medium is the selective agent inhibiting bacterial growth.

The choice of a suitable medium for enumeration of yeasts and molds from food samples is greatly dependent on the nature of the foodstuff under investigation and the organisms that occur on them. Yeast Extract Glucose Chloramphenicol Agar remains bacteriostatic when inoculated with not greater than 1mL of a 10-1 dilution of food sample and subsequently incubated for not greater than five days at 25°C as is the customary practice in food mycology.
TRUSTED_SUSTAINABILITY

Specifications

Color Ivory, transparent
Content And Storage 6°C to 12°C
Description Yeast Extract Glucose Chloramphenicol Medium
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Microbiology Media
Fill Volume 17 mL
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.

For laboratory use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.