missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Yeast Extract Agar

Product Code. 12962618
Change view
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12962618 10 Plates Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12962618 Supplier Thermo Scientific™ Supplier No. PO0441A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Support robust microbial growth with Yeast Extract Agar.

Yeast Extract Agar is ideal for cultivating a wide range of bacteria and fungi in laboratory settings.

TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Content And Storage 2°C to 12°C
Format 90 mm
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Volume 17 mL
Quantity 10 Plates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.