Learn More
Chemsplash Xtreme 50 SMS T5/6 Disposable Coverall Red
GREENER_CHOICE
Product Code. 30058323
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30058323 Small Pack of 25
30058373 Medium Pack of 25
30058375 Large Pack of 25
30058318 X-Large Pack of 25
30058335 2X-Large Pack of 25
30058271 3X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30058323 Supplier Chemsplash Supplier No. 2544.500.004
Add to basket
Add to basket
Xtreme SMS 50 disposable coveralls featuring an elasticated hood, cuffs and ankles, with a two way zip and an adhesive cover flap.
Chemsplash Xtreme SMS 50 Coveralls
  • 50GSM Breathable SMS Fabric
  • Elasticated 3 Piece Hood
  • Elasticated Cuffs and Ankles
  • Two Way Zip with Adhesive Zip Flap
  • Silicone and Latex Free
  • Comes without the chest label
  • PFAS Free
  • See Style 2618 for White Sizes 4XL-5XL-6XL
Description: Chemsplash Xtreme SMS 50 disposable coveralls are made from extremely breathable SMS fabric suitable for prolonged periods of use. Featuring an elasticated hood, cuffs, and ankles, with a two-way zip and an adhesive cover flap. Its strong and robust properties provide a good chemical barrier, especially to dry hazardous particles, whilst being extremely comfortable to wear. Offering the best value within our Cat III Type 5 and 6 range. Available in White, Blue, Red, and Orange.
Specifications
Product Type Hooded Coveralls
Size Small
Gender Unisex
Material SMS Fabric
Color Red
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Stitched
Sterility Non-sterile
Resistance Particle
Latex Free Yes
No. of Pockets None