Learn More
Chemsplash Xtreme 50 SMS T5/6 Disposable Coverall Red
GREENER_CHOICE
Product Code. 30058318
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30058318 X-Large Pack of 25
30058323 Small Pack of 25
30058373 Medium Pack of 25
30058375 Large Pack of 25
30058335 2X-Large Pack of 25
30058271 3X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30058318 Supplier Chemsplash Supplier No. 2544.500.007
Add to basket
Edge
Add to basket
Xtreme SMS 50 disposable coveralls featuring an elasticated hood, cuffs and ankles, with a two way zip and an adhesive cover flap.
Chemsplash Xtreme SMS 50 Coveralls
  • 50GSM Breathable SMS Fabric
  • Elasticated 3 Piece Hood
  • Elasticated Cuffs and Ankles
  • Two Way Zip with Adhesive Zip Flap
  • Silicone and Latex Free
  • Comes without the chest label
  • PFAS Free
  • See Style 2618 for White Sizes 4XL-5XL-6XL
Description: Chemsplash Xtreme SMS 50 disposable coveralls are made from extremely breathable SMS fabric suitable for prolonged periods of use. Featuring an elasticated hood, cuffs, and ankles, with a two-way zip and an adhesive cover flap. Its strong and robust properties provide a good chemical barrier, especially to dry hazardous particles, whilst being extremely comfortable to wear. Offering the best value within our Cat III Type 5 and 6 range. Available in White, Blue, Red, and Orange.
Specifications
Product Type Hooded Coveralls
Size X-Large
Gender Unisex
Material SMS Fabric
Color Red
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Stitched
Sterility Non-sterile
Resistance Particle
Latex Free Yes
No. of Pockets None