Learn More
Waters Corporation XSelect CSH Prep C18 Column
XSelect CSH C18 columns can be used as general purpose columns that provide alternative selectivity compared to other C18 sorbents.
£2265.00 - £18900.00
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Particle Size | 5 μm |
Pore Size | 130 Å |
Quantity | 1/Ea. |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
17173132
|
Waters Corporation
186005497 |
50 mm | 250 mm |
£18900.00
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
17113112
|
Waters Corporation
186005423 |
30 mm | 50 mm |
£4650.00
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
17153132
|
Waters Corporation
186005495 |
50 mm | 100 mm |
£11910.00
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
17163132
|
Waters Corporation
186005496 |
50 mm | 150 mm |
£15000.00
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
17143132
|
Waters Corporation
186005494 |
50 mm | 50 mm |
£8715.00
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
17183102
|
Waters Corporation
186005420 |
19 mm | 50 mm |
£2265.00
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
17133132
|
Waters Corporation
186005493 |
30 mm | 250 mm |
£12135.00
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
17193102
|
Waters Corporation
186005421 |
19 mm | 100 mm |
£3735.00
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
17103112
|
Waters Corporation
186005422 |
19 mm | 150 mm |
£4470.00
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
10558720
|
Waters Corporation
186005492 |
19 mm | 250 mm |
£5745.00
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
10019479
|
Waters Corporation
186005425 |
30 mm | 100 mm |
£6870.00
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
17123112
|
Waters Corporation
186005424 |
30 mm | 75 mm |
£5925.00
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
17133112
|
Waters Corporation
186005426 |
30 mm | 150 mm |
£8610.00
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
Description
- Charged Surface Hybrid (CSH) technology helps you overcome column related problems such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
Specifications
6000 psi (415 Bar) | |
5 μm | |
1/Ea. | |
Yes |
Spherical | |
130 Å | |
L1 | |
Column |