missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation XSelect CSH Prep C18 Column

XSelect CSH C18 columns can be used as general purpose columns that provide alternative selectivity compared to other C18 sorbents.

Brand:  Waters Corporation 186005494

Additional Details : Weight : 3.00050kg

 View more versions of this product

Product Code. 17143132

  • £8715.00 / Each
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

  • Charged Surface Hybrid (CSH) technology helps you overcome column related problems such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
Specifications

Specifications

50 mm
6000 psi (415 Bar)
5 μm
1/Ea.
Yes
50 mm
Spherical
130 Å
L1
Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers