missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XRCC2 Recombinant Rabbit Monoclonal Antibody (1T9D5)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558007
This item is not returnable.
View return policy
Descrizione
Immunogen sequence: MCSAFHRAES GTELLARLEG RSSLKEIEPN LFADEDSPVH GDILEFHGPE GTGKTEMLYH LTARCILPKS EGGLEVEVLF IDT.
The x-ray repair cross-complementing (XRCC) proteins are responsible for efficiently repairing and maintaining genetic stability following DNA base damage. These genes share sequence similarity with the yeast DNA repair protein Rad5. XRCC2 and XRCC3 are both involved in maintaining chromosome stability during cell division. XRCC2 is required for efficient repair of DNA double-strand breaks by homologous recombination between sister chromatids, and XRCC3 interacts directly with Rad51 to cooperate with Rad51 during recombinational repair.
Specifica
| XRCC2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O43543, Q9CX47 | |
| XRCC2 | |
| Recombinant protein (or fragment) | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1T9D5 | |
| Unconjugated | |
| XRCC2 | |
| 4921524O04Rik; 8030409M04Rik; DKFZp781P0919; DNA repair protein XRCC2; FANCU; RAD51; RecA; recA/RAD51 family protein; RGD1564823; X-ray repair complementing defective repair in Chinese hamster cells 2; X-ray repair cross complementing 2; X-ray repair cross complementing protein 2; X-ray repair cross-complementing protein 2; Xrcc2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 499966, 57434, 7516 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto