missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XRCC2 Recombinant Rabbit Monoclonal Antibody (1T9D5)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | XRCC2 |
|---|---|
| Clone | 1T9D5 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30283504
|
Invitrogen™
MA558007 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30284723
![]() |
Invitrogen™
MA558008 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: MCSAFHRAES GTELLARLEG RSSLKEIEPN LFADEDSPVH GDILEFHGPE GTGKTEMLYH LTARCILPKS EGGLEVEVLF IDT.
The x-ray repair cross-complementing (XRCC) proteins are responsible for efficiently repairing and maintaining genetic stability following DNA base damage. These genes share sequence similarity with the yeast DNA repair protein Rad5. XRCC2 and XRCC3 are both involved in maintaining chromosome stability during cell division. XRCC2 is required for efficient repair of DNA double-strand breaks by homologous recombination between sister chromatids, and XRCC3 interacts directly with Rad51 to cooperate with Rad51 during recombinational repair.Specifications
| XRCC2 | |
| 1 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O43543, Q9CX47 | |
| 499966, 57434, 7516 | |
| Recombinant protein (or fragment) | |
| Antibody |
| 1T9D5 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| XRCC2 | |
| 4921524O04Rik; 8030409M04Rik; DKFZp781P0919; DNA repair protein XRCC2; FANCU; RAD51; RecA; recA/RAD51 family protein; RGD1564823; X-ray repair complementing defective repair in Chinese hamster cells 2; X-ray repair cross complementing 2; X-ray repair cross complementing protein 2; X-ray repair cross-complementing protein 2; Xrcc2 | |
| XRCC2 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title