missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XRCC2 Mouse anti-Human, Clone: 3D9A1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | XRCC2 |
|---|---|
| Clone | 3D9A1 |
| Concentration | 1 mg/mL |
| Applications | Immunofluorescence, Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16808735
|
Proteintech
66652-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898725
|
Proteintech
66652-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The x-ray repair cross-complementing (XRCC) proteins are responsible for efficiently repairing and maintaining genetic stability following DNA base damage. These genes share sequence similarity with the yeast DNA repair protein Rad5. XRCC2 and XRCC3 are both involved in maintaining chromosome stability during cell division. XRCC2 is required for efficient repair of DNA double-strand breaks by homologous recombination between sister chromatids, and XRCC3 interacts directly with Rad51 to cooperate with Rad51 during recombinational repair.Specifications
| XRCC2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DKFZp781P0919, DNA repair protein XRCC2, XRCC2 | |
| XRCC2 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 3D9A1 | |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O43543 | |
| 7516 | |
| XRCC2 Fusion Protein Ag14421 | |
| Primary | |
| -20°C | |
| XRCC2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title