missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XRCC2 Mouse anti-Human, Clone: 3D9A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66652-1-IG-20UL
This item is not returnable.
View return policy
Description
The x-ray repair cross-complementing (XRCC) proteins are responsible for efficiently repairing and maintaining genetic stability following DNA base damage. These genes share sequence similarity with the yeast DNA repair protein Rad5. XRCC2 and XRCC3 are both involved in maintaining chromosome stability during cell division. XRCC2 is required for efficient repair of DNA double-strand breaks by homologous recombination between sister chromatids, and XRCC3 interacts directly with Rad51 to cooperate with Rad51 during recombinational repair.Specifications
| XRCC2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43543 | |
| XRCC2 | |
| XRCC2 Fusion Protein Ag14421 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 3D9A1 | |
| Unconjugated | |
| XRCC2 | |
| DKFZp781P0919, DNA repair protein XRCC2, XRCC2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 7516 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction