missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Xplorer™ BASIC - Multichannel Trade-In Pipette
GREENER_CHOICE

Product Code. p-200132153 Shop All Eppendorf Products
Click to view available options
Number of Channels:
8
12
Color:
Green
Medium Gray
Orange
Yellow
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15260765

Brand: Eppendorf™ 4861000570

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Eliminate potential error risks associated with manual pipetting with pipettes

  • Electronic single, 8 and 12 channel pipettes designed to aspirate and dispense liquid volumes from 0.005 to 10 mL
  • Intuitive operating concept and ergonomic design
  • Multifunctional rocker
  • Selection dial
  • Innovative ejector
  • Ergonomic display angle, Convenient handle
  • Powerful rechargeable battery
  • Flexible charging concept
  • Spring-loaded tip cone
  • Adjustment to external conditions (geographic height, liquids with high density or similar factors)
TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips epT.I.P.S.
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Medium Gray
Disposable Reusable
For Use With epT.I.P.S. Pipette Tip (10 μL)
Grip Type Ergonomic
Material Plastic
Number of Channels 8
Programmability Step-based
Sterility Non-sterile
Type Multichannel Pipette
Volume (Metric) 0.5 to 10 μL
For Use With (Application) Pipetting and mixing
For Use With (Equipment) Eppendorf Xplorer Pipettes
Quantity 1 Each
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.