missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£143.00 - £251.00
Specifications
| Antigen | XPC |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:500-1:3000 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622589
|
Novus Biologicals
NBP3-12570-100ul |
100 μg |
£251.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671909
|
Novus Biologicals
NBP3-12570-50ul |
50 μg |
£143.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
XPC Polyclonal antibody specifically detects XPC in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| XPC | |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| PBS, pH7.0 | |
| 7508 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:500-1:3000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| complementation group C, p125, RAD4, Xeroderma pigmentosum group C-complementing protein, xeroderma pigmentosum, complementation group C, XP3, XPCCDNA repair protein complementing XP-C cells | |
| Produced in rabbits immunized with E. coli-derived Human XPC fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title