missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12570-100ul
This item is not returnable.
View return policy
Description
XPC Polyclonal antibody specifically detects XPC in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| XPC | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:500-1:3000 | |
| complementation group C, p125, RAD4, Xeroderma pigmentosum group C-complementing protein, xeroderma pigmentosum, complementation group C, XP3, XPCCDNA repair protein complementing XP-C cells | |
| Produced in rabbits immunized with E. coli-derived Human XPC fragment | |
| 100 μg | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 7508 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction