missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ XLD Agar / Brilliance™ Salmonella Bi-plate

Product Code. 11984172
Change view
Click to view available options
Format:
90mm biplate
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size
11984172 90mm biplate Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11984172 Supplier Thermo Scientific™ Supplier No. PO5248E
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Work efficiently and according to the ISO method for Salmonella detection with a bi-plate that combines both Thermo Scientific™ XLD Agar and Brilliance™ Salmonella.

Work efficiently and according to the ISO method for Salmonella detection with a bi-plate that combines both Thermo Scientific™ XLD Agar and Brilliance™ Salmonella.

Read more results from a single patient sample and produce more diagnostic details of pathogenic organisms while reducing storage capacity issues in the incubator with a bi-plate that includes two widely used microbiological media.

XLD Agar is a selective media for the isolation and differentiation of Salmonella, relying on xylose fermentation, lysine decarboxylation and production of hydrogen sulfide for the primary differentiation of Shigella and Salmonella spp. from non-pathogenic bacteria. The sensitivity and selectivity of XLD Agar exceeds that of traditional plating media, e.g., Eosin Methylene Blue and Bismuth Sulphite agars, which tend to suppress the growth of shigellae. The recovery of salmonellae and shigellae is not obscured by profuse growth of other species; therefore, XLD Agar is ideal for the screening of samples containing mixed flora and suspected of harbouring enteric pathogens, e.g., medical specimens or food products. XLD Agar is recommended in the ISO 6579 testing of Salmonella and is tested according to ISO 11133-2.

Brilliance Salmonella Agar provides a highly selective and easy-to-read medium for the presumptive identification of Salmonella from food and environmental samples. The Inhibigen contained in this medium specifically targets Escherichia coli, with additional compounds added to suppress the growth of other non-target organisms.

Differentiation of Salmonella from other organisms is achieved through the inclusion of two chromogens that target specific enzymes: caprylate esterase and ß-glucosidase. The action of the enzymes on the chromogens result in a build-up of color within the colony, with the color produced depends on which enzymes the organism possesses. The action of caprylate esterase, present in all salmonellae, results in a purple colony.

Some other Enterobacteriaceae species also produce caprylate esterase, but these are either inhibited or differentiated from Salmonella by the ß-glucosidase substrate. This results in blue colonies which are easy to distinguish from the purple Salmonella colonies.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color XLD: Vermilion, transparent. SALM: Cream to pink tinged, opaque
Content And Storage 2°C to 12°C
Description XLD Agar / Brilliance Salmonella Bi-plate
Format 90mm biplate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 14g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.