missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XIRP1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £357.00
Specifications
| Antigen | XIRP1 |
|---|---|
| Dilution | Western Blot 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18349842
|
Novus Biologicals
NBP3-11953-25UL |
25 μg |
£145.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18302862
|
Novus Biologicals
NBP3-11953-100UL |
100 μg |
£357.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
XIRP1 Polyclonal antibody specifically detects XIRP1 in Mouse samples. It is validated for Western BlotSpecifications
| XIRP1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| cardiomyopathy associated 1, CMYA1, DKFZp779C1255, xin actin-binding repeat containing 1 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser295 conjugated to KLH. (UniProt: O70373) | |
| Antigen Affinity-purified |
| Western Blot 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 100 ul in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. | |
| 165904 | |
| Primary | |
| Store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title