missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XIRP1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-11953-100UL
This item is not returnable.
View return policy
Description
XIRP1 Polyclonal antibody specifically detects XIRP1 in Mouse samples. It is validated for Western Blot
Specifications
| XIRP1 | |
| Polyclonal | |
| Western Blot 1:1000 | |
| cardiomyopathy associated 1, CMYA1, DKFZp779C1255, xin actin-binding repeat containing 1 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser295 conjugated to KLH. (UniProt: O70373) | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 100 ul in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 165904 | |
| Store at -20C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction