missing translation for 'onlineSavingsMsg'
Learn More
Learn More
xCT Antibody (23H6), Novus Biologicals™
Human Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | xCT |
|---|---|
| Clone | 23H6 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30023486
|
Novus Biologicals
NBP3-26315-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30029365
|
Novus Biologicals
NBP3-26315-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
xCT Monoclonal antibody specifically detects xCT in Human samples. It is validated for ELISASpecifications
| xCT | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Amino acid transport system xc-, Calcium channel blocker resistance protein CCBR1, CCBR1, cystine/glutamate transporter, SLC7A11, solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11, Solute carrier family 7 member 11, solute carrier family 7, (cationic amino acid transporter, y+ system) member 11, xCT | |
| Recombinant Human xCT protein [UniProt Q9UPY5] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 23H6 | |
| ELISA | |
| Unconjugated | |
| Human | |
| Amino Acids Drugs and other small molecules, Cancer, Cell Biology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 23657 | |
| IgG1 | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title