missing translation for 'onlineSavingsMsg'
Learn More
Learn More
xCT Antibody (23H6), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-26315-50ul
This item is not returnable.
View return policy
Description
xCT Monoclonal antibody specifically detects xCT in Human samples. It is validated for ELISA
Specifications
| xCT | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| Human | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 23H6 | |
| ELISA | |
| Amino acid transport system xc-, Calcium channel blocker resistance protein CCBR1, CCBR1, cystine/glutamate transporter, SLC7A11, solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11, Solute carrier family 7 member 11, solute carrier family 7, (cationic amino acid transporter, y+ system) member 11, xCT | |
| Recombinant Human xCT protein [UniProt Q9UPY5] | |
| 50 μL | |
| Amino Acids Drugs and other small molecules, Cancer, Cell Biology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
| 23657 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction