Learn More
Fisherbrand™ Top-Line Pipette Filter Tips
Eliminate sample and pipette contamination. Fisherbrand™ Top-Line Pipette Filter Tips prevent impurities from passing through micropore filter.
Brand:  Fisherbrand™ FI2-113-96-0
 View more versions of this product
Product Code. 10102512
  • £60.63 / Pack of 960
Estimated Shipment: 10-10-2024
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Filters are made of high-quality hydrophobic virgin polyethylene without additives
  • Filters block aerosols and liquids through micropore filter technology
  • Optimized cone geometry for a secure fit
  • Graduation marks on tip at standard volumes (except 0.5-10/20µl filter tip)
  • Conical tip outlet prevents drop formation at the outlet opening of tip
  • Tips manufactured from autoclavable, non-wettable polypropylene
  • Sterile packs available (beta irradiation)
Specifications
Specifications
1 to 200 μL
Sterile
Autoclavable
Beta radiation
Yes
Standard Volumes
Filtered
Clear
50.8mm
Rack
Virgin Polypropylene