missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MRS Agar

Product Code. 12854142
Change view
Click to view available options
Fill Weight:
200-204 g
200.0-204.0g
250 to 254 g
Format:
250 mL sirop bottle
250mL sirop bottle
300 mL Sirop Bottle
Unit Size:
Pack of 10
Pack of 250
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Format unitSize
12854142 250 to 254 g 300 mL Sirop Bottle Pack of 250
13245469 200-204 g 250 mL sirop bottle Pack of 10
15304885 200.0-204.0g 250mL sirop bottle Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12854142 Supplier Thermo Scientific™ Supplier No. BO0969T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use this agar for the cultivation of lactobacilli

The formula of MRS Agar is based on that of de Man, Rogosa and Sharpe.

TRUSTED_SUSTAINABILITY

Specifications

Color Amber
Content And Storage 2° to 25°C
Format 300 mL Sirop Bottle
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 250 to 254 g
Packaging Type 10 Sirop Bottles with screw cap closures in a box
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.