missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sabouraud Dextrose Agar (Slope)

Product Code. 12736949
Click to view available options
Quantity:
24 x 10 mL
Unit Size:
Pack of 24
This item is not returnable. View return policy

Product Code. 12736949

Brand: Thermo Scientific™ BO0342E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SABOURAUD DEXTROSE AGAR (SLOPE) 24 x 10 mL: For the cultivation of yeast and fungi.

  • SABOURAUD DEXTROSE AGAR (SLOPE) 24 x 10 mL: For the cultivation of yeast and fungi
TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2 to 25°C
Format Universal bottle
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 10.0-10.3g
Packaging Type Twenty-four universal bottles with screw cap closures in a box.
Quantity 24 x 10 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.