missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Buffered Nitrate Motility Medium
Click to view available options
Fill Weight:
10.0 to 10.3 g
Unit Size:
Pack of 24
Description
Determine bacterial motility and nitrate reduction ability, aiding in the identification of enteric and other motile bacteria using Buffered Nitrate Motility Medium.
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2°C to 10°C |
| Format | Universal bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Microbiology Media |
| Fill Weight | 10.0 to 10.3 g |
| Packaging Type | 24 Universal Bottles with plastic screw cap closures in a box |
| Quantity | 24/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction