missing translation for 'onlineSavingsMsg'
Learn More

IKA™ X20 BMT-20-G-M-gamma Tube with Stirring Element

Product Code. 16208531 Shop All IKA Products
Change view
Click to view available options
For Use With (Equipment):
ULTRA-TURRAX™
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
16208531 ULTRA-TURRAX™ Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16208531 Supplier IKA™ Supplier No. 0020017448
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

BMT-20-G-M-gamma Tube for grinding with glass balls.

  • Components: Tube, glas balls, cover
  • Dry milling of dry and brittle samples (e.g. kaolin, gypsum, coloured pigments, tablets), cell maceration, processing of materials mixed with fluids
  • Can be used for sampling during operation, sample preparation under sterile conditions, and germ free conservation of samples (tissue, blood, etc.)
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2 to 15 mL
For Use With (Equipment) ULTRA-TURRAX™
Packaging Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.