missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Tryptone Soya Broth (Bottled)

Product Code. 15632129
Click to view available options
Fill Weight:
10.01 to 0.3 g
10.0g + 0.3g
100 to 101.5 g
100.0 - 101.5g
100.0g + 1.5 g
1000 to 1005 g
20.0 - 20.3 g
450 to 454 g
500.0 - 504.0 g
90 to 90.5 g
Unit Size:
1 set
Pack of 10
Pack of 24
This item is not returnable. View return policy

Product Code. 15632129

Brand: Thermo Scientific™ BO0282Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Grow a wide variety of microprganisms using this medium in convenient ready prepared bottles.

Grow a wide variety of microorganisms with Thermo Scientific™ Tryptone Soya Broth ready-prepared bottled medium. The general purpose medium, containing two peptones, is suitable for the cultivation of both aerobes and anaerobes. It is specified in International Pharmacopoeia1,2,3,4 and food testing methods for the growth of a wide variety of organisms.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready-to-use bottles of Tryptone Soya Broth.

  • Conforms to formulations detailed in International Pharmacopoeia.
  • Convenient: pre-filled bottles are ready for immediate use.
  • Simple: easy to use with minimum training.
  • Long shelf life: up to 12 months.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Thermo Scientific™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw, clear
Content And Storage 2°C to 25°C
Format 500 mL DIN bottle
Manufacturing Origin United Kingdom
Product Type Microbiology Media
Fill Weight 450 to 454 g
Packaging Type 10 DIN bottles with injectable closures in a box.
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.