missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptone Soya Broth

Product Code. 12930109
Click to view available options
Fill Weight:
90-91.5 g
Format:
125 mL sirop bottle
Quantity:
10 x 90 mL
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12930109

Brand: Thermo Scientific™ BO0479Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A non-selective broth with neutralizers.

Tryptone Soya Broth is a non-selective broth with neutralizers.

  • TRYPTONE SOYA BROTH + 0.3% LECITHIN + 2% Tween™ 10 x 90 mL
TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2 to 25°C
Format 125 mL sirop bottle
Manufacturing Origin United Kingdom
Product Type Broth
Fill Weight 90-91.5 g
Packaging Type 10 sirop bottles with screw cap closures in a box.
Quantity 10 x 90 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.